Anti-USP13 Mouse Monoclonal Antibody [clone: 4G1]

Toimittaja: OriGene
ORIGCF500327EA 749 EUR
ORIGCF500327
Anti-USP13 Mouse Monoclonal Antibody [clone: 4G1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: USP13
Clonality: Monoclonal
Clone: 4G1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR