Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 3C6]

Toimittaja: OriGene
ORIGCF502302EA 749 EUR
ORIGCF502302
Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 3C6]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: CTNNB1
Clonality: Monoclonal
Clone: 3C6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR