Anti-RDH14 Mouse Monoclonal Antibody [clone: 6C3]

Toimittaja: OriGene
ORIGCF502315EA 749 EUR
ORIGCF502315
Anti-RDH14 Mouse Monoclonal Antibody [clone: 6C3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-500

Type: Primary
Antigen: RDH14
Clonality: Monoclonal
Clone: 6C3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR