Anti-SERPINB4 Mouse Monoclonal Antibody [clone: 3C5]

Toimittaja: OriGene
ORIGCF502325EA 749 EUR
ORIGCF502325
Anti-SERPINB4 Mouse Monoclonal Antibody [clone: 3C5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SERPINB4
Clonality: Monoclonal
Clone: 3C5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR