Anti-GBP2 Mouse Monoclonal Antibody [clone: 5E10]

Toimittaja: OriGene
ORIGCF500669EA 770 EUR
ORIGCF500669
Anti-GBP2 Mouse Monoclonal Antibody [clone: 5E10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50

Type: Primary
Antigen: GBP2
Clonality: Monoclonal
Clone: 5E10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR