Anti-CD5 Mouse Monoclonal Antibody [clone: 6F7]

Toimittaja: OriGene
ORIGCF501334EA 749 EUR
ORIGCF501334
Anti-CD5 Mouse Monoclonal Antibody [clone: 6F7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 6F7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR