Anti-TTC32 Mouse Monoclonal Antibody [clone: 1F4]

Toimittaja: OriGene
ORIGCF501340EA 770 EUR
ORIGCF501340
Anti-TTC32 Mouse Monoclonal Antibody [clone: 1F4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TTC32
Clonality: Monoclonal
Clone: 1F4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR