Anti-CYB5R3 Mouse Monoclonal Antibody [clone: 2A10]

Toimittaja: OriGene
ORIGCF501353EA 749 EUR
ORIGCF501353
Anti-CYB5R3 Mouse Monoclonal Antibody [clone: 2A10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: CYB5R3
Clonality: Monoclonal
Clone: 2A10
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR