Anti-NEUROG1 Mouse Monoclonal Antibody [clone: 8G8]

Toimittaja: OriGene
ORIGCF500348EA 749 EUR
ORIGCF500348
Anti-NEUROG1 Mouse Monoclonal Antibody [clone: 8G8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1000; Immunofluorescence: 1:50; Immunoprecipitation: 2-4?g/mg

Type: Primary
Antigen: NEUROG1
Clonality: Monoclonal
Clone: 8G8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR