Anti-AK1 Mouse Monoclonal Antibody [clone: 4A1]

Toimittaja: OriGene
ORIGCF500361EA 770 EUR
ORIGCF500361
Anti-AK1 Mouse Monoclonal Antibody [clone: 4A1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: AK1
Clonality: Monoclonal
Clone: 4A1
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR