Anti-AK1 Mouse Monoclonal Antibody [clone: 5G8]
Toimittaja: OriGene
ORIGCF500362EA 770 EUR
ORIGCF500362
Anti-AK1 Mouse Monoclonal Antibody [clone: 5G8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000-2000; Immunofluorescence: 1:50
Type: Primary
Antigen: AK1
Clonality: Monoclonal
Clone: 5G8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human