Anti-SERPINA1 Mouse Monoclonal Antibody [clone: 11G2]

Toimittaja: OriGene
ORIGCF500378EA 749 EUR
ORIGCF500378
Anti-SERPINA1 Mouse Monoclonal Antibody [clone: 11G2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: SERPINA1
Clonality: Monoclonal
Clone: 11G2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR