Anti-AQP1 Mouse Monoclonal Antibody [clone: 10C11]

Toimittaja: OriGene
ORIGCF502356EA 749 EUR
ORIGCF502356
Anti-AQP1 Mouse Monoclonal Antibody [clone: 10C11]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: AQP1
Clonality: Monoclonal
Clone: 10C11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR