Anti-RNF144B Mouse Monoclonal Antibody [clone: 10A12]

Toimittaja: OriGene
ORIGCF500706EA 749 EUR
ORIGCF500706
Anti-RNF144B Mouse Monoclonal Antibody [clone: 10A12]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:100

Type: Primary
Antigen: RNF144B
Clonality: Monoclonal
Clone: 10A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR