Anti-ID3 Mouse Monoclonal Antibody [clone: 4D1]

Toimittaja: OriGene
ORIGCF500715EA 749 EUR
ORIGCF500715
Anti-ID3 Mouse Monoclonal Antibody [clone: 4D1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ID3
Clonality: Monoclonal
Clone: 4D1
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR