Anti-PRL Mouse Monoclonal Antibody [clone: 9D12]

Toimittaja: OriGene
ORIGCF500718EA 770 EUR
ORIGCF500718
Anti-PRL Mouse Monoclonal Antibody [clone: 9D12]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: PRL
Clonality: Monoclonal
Clone: 9D12
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR