Anti-IDH3A Mouse Monoclonal Antibody [clone: 5D2]

Toimittaja: OriGene
ORIGCF500736EA 749 EUR
ORIGCF500736
Anti-IDH3A Mouse Monoclonal Antibody [clone: 5D2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: IDH3A
Clonality: Monoclonal
Clone: 5D2
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR