Anti-AKR1A1 Mouse Monoclonal Antibody [clone: 8E12]

Toimittaja: OriGene
ORIGCF500741EA 749 EUR
ORIGCF500741
Anti-AKR1A1 Mouse Monoclonal Antibody [clone: 8E12]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Flow Cytometry: 1:100

Type: Primary
Antigen: AKR1A1
Clonality: Monoclonal
Clone: 8E12
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR