Anti-COX6A1 Mouse Monoclonal Antibody [clone: 5B6]

Toimittaja: OriGene
ORIGCF501395EA 749 EUR
ORIGCF501395
Anti-COX6A1 Mouse Monoclonal Antibody [clone: 5B6]
Vasta-aineet
Recommended Dilutions: Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: COX6A1
Clonality: Monoclonal
Clone: 5B6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR