Anti-TMEM80 Mouse Monoclonal Antibody [clone: 3B6]

Toimittaja: OriGene
ORIGCF501408EA 749 EUR
ORIGCF501408
Anti-TMEM80 Mouse Monoclonal Antibody [clone: 3B6]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TMEM80
Clonality: Monoclonal
Clone: 3B6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR