Anti-MAP2K4 Mouse Monoclonal Antibody [clone: 8A8]
Toimittaja: OriGene
ORIGCF500403EA 749 EUR
ORIGCF500403
Anti-MAP2K4 Mouse Monoclonal Antibody [clone: 8A8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:150; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l
Type: Primary
Antigen: MAP2K4
Clonality: Monoclonal
Clone: 8A8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human