Anti-SERPINB4 Mouse Monoclonal Antibody [clone: 4C8]

Toimittaja: OriGene
ORIGCF502386EA 749 EUR
ORIGCF502386
Anti-SERPINB4 Mouse Monoclonal Antibody [clone: 4C8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: SERPINB4
Clonality: Monoclonal
Clone: 4C8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR