Anti-EIF4E2 Mouse Monoclonal Antibody [clone: 1F5]

Toimittaja: OriGene
ORIGCF502402EA 749 EUR
ORIGCF502402
Anti-EIF4E2 Mouse Monoclonal Antibody [clone: 1F5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: EIF4E2
Clonality: Monoclonal
Clone: 1F5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR