Anti-PRKCE Mouse Monoclonal Antibody [clone: 1B4]
Toimittaja: OriGene
ORIGCF502417EA 749 EUR
ORIGCF502417
Anti-PRKCE Mouse Monoclonal Antibody [clone: 1B4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: PRKCE
Clonality: Monoclonal
Clone: 1B4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey