Anti-HDAC10 Mouse Monoclonal Antibody [clone: 6D4]

Toimittaja: OriGene
ORIGCF500754EA 770 EUR
ORIGCF500754
Anti-HDAC10 Mouse Monoclonal Antibody [clone: 6D4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunoprecipitation: 2-4?g/mg

Type: Primary
Antigen: HDAC10
Clonality: Monoclonal
Clone: 6D4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR