Anti-SORD Mouse Monoclonal Antibody [clone: 13G5]

Toimittaja: OriGene
ORIGCF500784EA 749 EUR
ORIGCF500784
Anti-SORD Mouse Monoclonal Antibody [clone: 13G5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50

Type: Primary
Antigen: SORD
Clonality: Monoclonal
Clone: 13G5
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR