Anti-IL1F6 Mouse Monoclonal Antibody [clone: 3A12]

Toimittaja: OriGene
ORIGCF501427EA 749 EUR
ORIGCF501427
Anti-IL1F6 Mouse Monoclonal Antibody [clone: 3A12]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: IL1F6
Clonality: Monoclonal
Clone: 3A12
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR