Anti-GCK Mouse Monoclonal Antibody [clone: 3E3]

Toimittaja: OriGene
ORIGCF500459EA 749 EUR
ORIGCF500459
Anti-GCK Mouse Monoclonal Antibody [clone: 3E3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:50

Type: Primary
Antigen: GCK
Clonality: Monoclonal
Clone: 3E3
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR