Anti-NRBP1 Mouse Monoclonal Antibody [clone: 4C1]

Toimittaja: OriGene
ORIGCF500461EA 749 EUR
ORIGCF500461
Anti-NRBP1 Mouse Monoclonal Antibody [clone: 4C1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: NRBP1
Clonality: Monoclonal
Clone: 4C1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR