Anti-SLC18A2 Mouse Monoclonal Antibody [clone: 10C11]

Toimittaja: OriGene
ORIGCF500463EA 749 EUR
ORIGCF500463
Anti-SLC18A2 Mouse Monoclonal Antibody [clone: 10C11]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50

Type: Primary
Antigen: SLC18A2
Clonality: Monoclonal
Clone: 10C11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR