Anti-MAPK1 Mouse Monoclonal Antibody [clone: 6F8]

Toimittaja: OriGene
ORIGCF500468EA 749 EUR
ORIGCF500468
Anti-MAPK1 Mouse Monoclonal Antibody [clone: 6F8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: MAPK1
Clonality: Monoclonal
Clone: 6F8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR