Anti-PRKCE Mouse Monoclonal Antibody [clone: 4B3]

Toimittaja: OriGene
ORIGCF502439EA 749 EUR
ORIGCF502439
Anti-PRKCE Mouse Monoclonal Antibody [clone: 4B3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PRKCE
Clonality: Monoclonal
Clone: 4B3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR