Anti-TTLL12 Mouse Monoclonal Antibody [clone: 4B2]

Toimittaja: OriGene
ORIGCF500789EA 770 EUR
ORIGCF500789
Anti-TTLL12 Mouse Monoclonal Antibody [clone: 4B2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: TTLL12
Clonality: Monoclonal
Clone: 4B2
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR