Anti-LEMD3 Mouse Monoclonal Antibody [clone: 3H2]

Toimittaja: OriGene
ORIGCF500795EA 770 EUR
ORIGCF500795
Anti-LEMD3 Mouse Monoclonal Antibody [clone: 3H2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Immunoprecipitation: 2-4?g/mg

Type: Primary
Antigen: LEMD3
Clonality: Monoclonal
Clone: 3H2
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR