Anti-EFNA2 Mouse Monoclonal Antibody [clone: 2D6]
Toimittaja: OriGene
ORIGCF501456EA 749 EUR
ORIGCF501456
Anti-EFNA2 Mouse Monoclonal Antibody [clone: 2D6]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: EFNA2
Clonality: Monoclonal
Clone: 2D6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human