Anti-DCX Mouse Monoclonal Antibody [clone: 4A3]

Toimittaja: OriGene
ORIGCF501778EA 749 EUR
ORIGCF501778
Anti-DCX Mouse Monoclonal Antibody [clone: 4A3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: DCX
Clonality: Monoclonal
Clone: 4A3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR