Anti-GAS7 Mouse Monoclonal Antibody [clone: 2D11]

Toimittaja: OriGene
ORIGCF501779EA 770 EUR
ORIGCF501779
Anti-GAS7 Mouse Monoclonal Antibody [clone: 2D11]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: GAS7
Clonality: Monoclonal
Clone: 2D11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR