Anti-AFP Mouse Monoclonal Antibody [clone: 4D2]

Toimittaja: OriGene
ORIGCF501780EA 749 EUR
ORIGCF501780
Anti-AFP Mouse Monoclonal Antibody [clone: 4D2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: 4D2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR