Anti-AFP Mouse Monoclonal Antibody [clone: 3B5]
Toimittaja: OriGene
ORIGCF501787EA 749 EUR
ORIGCF501787
Anti-AFP Mouse Monoclonal Antibody [clone: 3B5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100
Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: 3B5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human