Anti-EPM2AIP1 Mouse Monoclonal Antibody [clone: 2A2]

Toimittaja: OriGene
ORIGCF501790EA 749 EUR
ORIGCF501790
Anti-EPM2AIP1 Mouse Monoclonal Antibody [clone: 2A2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: EPM2AIP1
Clonality: Monoclonal
Clone: 2A2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR