Anti-STK39 Mouse Monoclonal Antibody [clone: 2E10]

Toimittaja: OriGene
ORIGCF500497EA 749 EUR
ORIGCF500497
Anti-STK39 Mouse Monoclonal Antibody [clone: 2E10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: STK39
Clonality: Monoclonal
Clone: 2E10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR