Anti-NUDT9 Mouse Monoclonal Antibody [clone: 5B5]

Toimittaja: OriGene
ORIGCF503275EA 786 EUR
ORIGCF503275
Anti-NUDT9 Mouse Monoclonal Antibody [clone: 5B5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: NUDT9
Clonality: Monoclonal
Clone: 5B5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR