Anti-ATG3 Mouse Monoclonal Antibody [clone: 3G3]

Toimittaja: OriGene
ORIGCF503299EA 749 EUR
ORIGCF503299
Anti-ATG3 Mouse Monoclonal Antibody [clone: 3G3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ATG3
Clonality: Monoclonal
Clone: 3G3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR