Anti-XPNPEP3 Mouse Monoclonal Antibody [clone: 5G2]

Toimittaja: OriGene
ORIGCF503304EA 749 EUR
ORIGCF503304
Anti-XPNPEP3 Mouse Monoclonal Antibody [clone: 5G2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: XPNPEP3
Clonality: Monoclonal
Clone: 5G2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR