Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 3C3]

Toimittaja: OriGene
ORIGCF505618EA 749 EUR
ORIGCF505618
Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 3C3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: MAP2K3
Clonality: Monoclonal
Clone: 3C3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR