Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 1D2]

Toimittaja: OriGene
ORIGCF505631EA 749 EUR
ORIGCF505631
Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 1D2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: MAP2K3
Clonality: Monoclonal
Clone: 1D2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR