Anti-TRAPPC4 Mouse Monoclonal Antibody [clone: 3A8]
Toimittaja: OriGene
ORIGCF505632EA 749 EUR
ORIGCF505632
Anti-TRAPPC4 Mouse Monoclonal Antibody [clone: 3A8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500
Type: Primary
Antigen: TRAPPC4
Clonality: Monoclonal
Clone: 3A8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human