Anti-GSTO2 Mouse Monoclonal Antibody [clone: 4G1]

Toimittaja: OriGene
ORIGCF503677EA 749 EUR
ORIGCF503677
Anti-GSTO2 Mouse Monoclonal Antibody [clone: 4G1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: GSTO2
Clonality: Monoclonal
Clone: 4G1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR