Anti-FAM119A Mouse Monoclonal Antibody [clone: 2G7]

Toimittaja: OriGene
ORIGCF503704EA 749 EUR
ORIGCF503704
Anti-FAM119A Mouse Monoclonal Antibody [clone: 2G7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: FAM119A
Clonality: Monoclonal
Clone: 2G7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR