Anti-PTPN7 Mouse Monoclonal Antibody [clone: 3B10]

Toimittaja: OriGene
ORIGCF503714EA 749 EUR
ORIGCF503714
Anti-PTPN7 Mouse Monoclonal Antibody [clone: 3B10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PTPN7
Clonality: Monoclonal
Clone: 3B10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR